Assay Details
| Gene Symbol: LOC501282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_576697.6 | XP_576697.3 | 2 - 3 | 241 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC501282| Gene Symbol: LOC501282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_576697.6 | XP_576697.3 | 2 - 3 | 241 | 59 | |
Set Membership: |
|