Assay Details
| Gene Symbol: Ttll11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001071863.5 | XP_001071863.3 | 7 - 8 | 2353 | 68 | |
| GenBank mRNA | BC169049.1 | - | 5 - 6 | 1183 | 7374372 | 68 |
More Information
Updated Mapping Information
Set Membership: |
|

Ttll11
Genomic Map