Assay Details
| Gene Symbol: Anapc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100532.1 | NP_001094002.1 | 11 - 12 | 2065 | 57 | |
| GenBank mRNA | BC107471.1 | - | 9 - 10 | 1169 | 57 | |
| BC166796.1 | - | 11 - 12 | 2065 | 7938000 | 57 | |
| FQ224461.1 | - | 11 - 12 | 2075 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anapc2
Genomic Map