Assay Details
| Gene Symbol: Apobec4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017492.1 | NP_001017492.1 | 1 - 2 | 161 | 73 | |
| GenBank mRNA | BC079272.1 | - | 1 - 2 | 161 | 7134003 | 73 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC079272.1, NM_001017492.1 transcripts.
Set Membership: |
|

Apobec4
Genomic Map