Assay Details
| Gene Symbol: Rgs18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047084.1 | NP_001040549.1 | 3 - 4 | 278 | 121 | |
| XM_008769548.1 | XP_008767770.1 | 3 - 4 | 304 | 121 | ||
| GenBank mRNA | AY651776.1 | - | 3 - 4 | 278 | 121 | |
More Information
Set Membership: |
|

Rgs18
Genomic Map