Assay Details
Gene Symbol: Malt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751798.3 | XP_003751846.1 | 2 - 3 | 613 | 61 | |
XM_006254888.2 | XP_006254950.1 | 2 - 3 | 613 | 61 | ||
XM_225927.8 | XP_225927.5 | 2 - 3 | 612 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
