Assay Details
Gene Symbol: Fam122c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257649.2 | XP_006257711.1 | 4 - 5 | 397 | 68 | |
XM_006257651.2 | XP_006257713.1 | 4 - 5 | 397 | 68 | ||
XM_008773640.1 | XP_008771862.1 | 4 - 5 | 397 | 68 | ||
XM_008773641.1 | XP_008771863.1 | 4 - 5 | 433 | 68 | ||
XM_008773642.1 | XP_008771864.1 | 4 - 5 | 397 | 68 | ||
XM_008773643.1 | XP_008771865.1 | 4 - 5 | 397 | 68 | ||
XM_008773644.1 | XP_008771866.1 | 4 - 5 | 397 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
