Assay Details
| Gene Symbol: Slc35d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106098.2 | NP_001099568.2 | 9 - 10 | 762 | 88 | |
| XM_006253493.2 | XP_006253555.1 | 9 - 10 | 1746 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc35d2
Genomic Map