Assay Details
Gene Symbol: RGD1305014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014025.1 | NP_001014047.1 | 1 - 2 | 168 | 87 | |
XM_008759885.1 | XP_008758107.1 | 1 - 2 | 1370 | 87 | ||
GenBank mRNA | BC083846.1 | - | 1 - 2 | 168 | 7121351 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
