Assay Details
| Gene Symbol: Dtx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047855.1 | NP_001041320.1 | 2 - 3 | 105 | 60 | |
| XM_006231114.2 | XP_006231176.1 | 3 - 4 | 1926 | 60 | ||
| GenBank mRNA | AY383692.1 | - | 2 - 3 | 105 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dtx4
Genomic Map