Assay Details
| Gene Symbol: LOC102546963 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760291.1 | XP_008758513.1 | 1 - 2 | 176 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008760291.1 transcripts.
Set Membership: |
|

LOC102546963
Genomic Map