Assay Details
| Gene Symbol: Suv420h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108512.1 | NP_001101982.1 | 1 - 2 | 316 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108512.1 transcripts.
Set Membership: |
|

Suv420h1
Genomic Map