Assay Details
| Gene Symbol: Suv420h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108512.1 | NP_001101982.1 | 4 - 5 | 1005 | 146 | |
| XM_008760150.1 | XP_008758372.1 | 2 - 3 | 387 | 146 | ||
| XM_008760151.1 | XP_008758373.1 | 3 - 4 | 689 | 146 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Suv420h1
Genomic Map