Assay Details
| Gene Symbol: Suv420h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108512.1 | NP_001101982.1 | 9 - 10 | 1536 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Suv420h1| Gene Symbol: Suv420h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108512.1 | NP_001101982.1 | 9 - 10 | 1536 | 88 | |
Set Membership: |
|