Assay Details
Gene Symbol: NEWGENE_1565505 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760350.1 | XP_008758572.1 | 3 - 4 | 1347 | 65 | |
XM_008760351.1 | XP_008758573.1 | 3 - 4 | 1716 | 65 | ||
XM_008760352.1 | XP_008758574.1 | 2 - 3 | 2332 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
