Assay Details
| Gene Symbol: Fam111a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109163.1 | NP_001102633.1 | 1 - 2 | 55 | 60 | |
| GenBank mRNA | FQ226103.1 | - | 1 - 2 | 54 | 60 | |
| FQ230484.1 | - | 1 - 2 | 53 | 60 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109163.1 transcripts.
Set Membership: |
|

Fam111a
Genomic Map