Assay Details
| Gene Symbol: Gfra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053398.1 | NP_445850.1 | 5 - 6 | 935 | 68 | |
| GenBank mRNA | AF184920.1 | - | 3 - 4 | 475 | 68 | |
| BC079378.1 | - | 5 - 6 | 935 | 7110848 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gfra3
Genomic Map