Assay Details
Gene Symbol: Mgrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013964.1 | NP_001013986.1 | 8 - 9 | 863 | 62 | |
XM_006245792.2 | XP_006245854.1 | 8 - 9 | 867 | 62 | ||
XM_006245793.2 | XP_006245855.1 | 8 - 9 | 864 | 62 | ||
XM_006245794.2 | XP_006245856.1 | 8 - 9 | 866 | 62 | ||
XM_006245795.2 | XP_006245857.1 | 8 - 9 | 863 | 62 | ||
XM_006245796.2 | XP_006245858.1 | 8 - 9 | 867 | 62 | ||
XM_008767485.1 | XP_008765707.1 | 8 - 9 | 863 | 62 | ||
XM_008767486.1 | XP_008765708.1 | 8 - 9 | 863 | 62 | ||
GenBank mRNA | BC083621.1 | - | 8 - 9 | 863 | 7134018 | 62 |
HE574489.1 | - | 8 - 9 | 727 | 62 | ||
HE574490.1 | - | 8 - 9 | 727 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
