Assay Details
| Gene Symbol: LOC100910567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749284.2 | XP_003749332.1 | 3 - 4 | 541 | 69 | |
| Gene Symbol: Aadacl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191706.1 | NP_001178635.1 | 429 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC100910567
Genomic Map