Assay Details
| Gene Symbol: Ufm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126080.2 | NP_001119552.1 | 3 - 4 | 194 | 65 | |
| GenBank mRNA | BC091395.1 | - | 3 - 4 | 130 | 7323172 | 65 |
| CB787557.1 | - | 3 - 4 | 232 | 65 | ||
| CV112030.1 | - | 2 - 3 | 84 | 65 | ||
| FQ229705.1 | - | 3 - 4 | 165 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ufm1
Genomic Map