Assay Details
| Gene Symbol: Eri3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006238735.2 | XP_006238797.1 | 3 - 4 | 714 | 71 | |
| XM_006238736.2 | XP_006238798.1 | 3 - 4 | 708 | 71 | ||
| XM_006238737.2 | XP_006238799.1 | 4 - 5 | 523 | 71 | ||
| XM_006238738.2 | XP_006238800.1 | 2 - 3 | 2765 | 71 | ||
| XM_006238739.1 | XP_006238801.1 | 3 - 4 | 714 | 71 | ||
| XM_006238741.2 | XP_006238803.1 | 3 - 4 | 410 | 71 | ||
| XM_008764085.1 | XP_008762307.1 | 3 - 4 | 2733 | 71 | ||
| XM_008764086.1 | XP_008762308.1 | 2 - 3 | 709 | 71 | ||
| XM_233435.6 | XP_233435.5 | 3 - 4 | 714 | 71 | ||
| GenBank mRNA | BC167080.1 | - | 2 - 3 | 354 | 71 | |
More Information
Set Membership: |
|

Eri3
Genomic Map