Assay Details
Gene Symbol: Galnt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172063.2 | NP_001165534.1 | 4 - 5 | 1052 | 53 | |
XM_006242294.2 | XP_006242356.1 | 6 - 7 | 2138 | 53 | ||
XM_006242295.2 | XP_006242357.1 | 6 - 7 | 1461 | 53 | ||
XM_006242296.2 | XP_006242358.1 | 6 - 7 | 1405 | 53 | ||
XM_008765651.1 | XP_008763873.1 | 7 - 8 | 1640 | 53 | ||
XM_008765652.1 | XP_008763874.1 | 6 - 7 | 1397 | 53 | ||
XM_008765653.1 | XP_008763875.1 | 6 - 7 | 1677 | 53 | ||
GenBank mRNA | FQ211776.1 | - | 6 - 7 | 1430 | 53 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
