Assay Details
| Gene Symbol: Elmod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109506.1 | NP_001102976.1 | 2 - 3 | 244 | 110 | |
| XM_006255302.2 | XP_006255364.1 | 2 - 3 | 491 | 110 | ||
| XM_006255303.2 | XP_006255365.1 | 2 - 3 | 290 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Elmod2
Genomic Map