Assay Details
Gene Symbol: Syce1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006255676.2 | XP_006255738.1 | 2 - 3 | 140 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
