Assay Details
| Gene Symbol: Ces2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106175.1 | NP_001099645.1 | 10 - 11 | 1474 | 94 | |
| XM_006255448.2 | XP_006255510.1 | 9 - 10 | 1222 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ces2g
Genomic Map