Assay Details
| Gene Symbol: Ces2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106175.1 | NP_001099645.1 | 6 - 7 | 961 | 63 | |
| XM_006255448.2 | XP_006255510.1 | 5 - 6 | 709 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ces2g
Genomic Map