Assay Details
Gene Symbol: Chst5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006255632.2 | XP_006255694.1 | 2 - 3 | 293 | 91 | |
GenBank mRNA | FQ212819.1 | - | 2 - 3 | 293 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006255632.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
