Assay Details
| Gene Symbol: Ddx19b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005895.2 | NP_001005895.2 | 2 - 3 | 252 | 86 | |
| GenBank mRNA | AY751756.1 | - | 2 - 3 | 95 | ||
| EV774940.1 | - | 2 - 3 | 252 | 86 | ||
| Gene Symbol: Ddx19a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY303540.1 | - | 95 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx19b
Genomic Map