Assay Details
| Gene Symbol: Ces2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044258.2 | NP_001037723.1 | 4 - 5 | 591 | 78 | |
| NM_001190346.1 | NP_001177275.1 | 4 - 5 | 591 | 78 | ||
| XM_008772523.1 | XP_008770745.1 | 4 - 5 | 610 | 78 | ||
| XM_008772524.1 | XP_008770746.1 | 5 - 6 | 733 | 78 | ||
| GenBank mRNA | BC107806.1 | - | 4 - 5 | 561 | 7368719 | 78 |
More Information
Updated Mapping Information
Set Membership: |
|

Ces2h
Genomic Map