Assay Details
Gene Symbol: Scara3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108870.1 | NP_001102340.1 | 3 - 4 | 462 | 93 | |
GenBank mRNA | FQ211593.1 | - | 3 - 4 | 479 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
