Assay Details
Gene Symbol: Scara3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108870.1 | NP_001102340.1 | 4 - 5 | 573 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
