Assay Details
| Gene Symbol: Ccdc129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191973.1 | NP_001178902.1 | 11 - 12 | 2627 | 79 | |
| XM_008762911.1 | XP_008761133.1 | 11 - 12 | 2644 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc129
Genomic Map