Assay Details
| Gene Symbol: Atp6v1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108318.2 | NP_001101788.1 | 2 - 3 | 186 | 83 | |
| XM_006248348.2 | XP_006248410.1 | 2 - 3 | 387 | 83 | ||
| XM_006248349.2 | XP_006248411.1 | 2 - 3 | 378 | 83 | ||
| GenBank mRNA | FQ211993.1 | - | 2 - 3 | 219 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P347007 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P11066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187280 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931314 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321403 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659280 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703599 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5280 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179901 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P347007 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11066 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11066 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P347007 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P347007 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P11066 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11066 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15246 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10751166 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751163 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751164 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823610 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751162 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751165 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10751161 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10823609 | - | - |
| Rat Genome 230 2.0 Array | 1382048_at | - | - |
| Rat Genome 230 2.0 Array | 1374431_at | - | - |
| Rat Genome 230 2.0 Array | 1382048_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374431_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1382048_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374431_at | - | - |
| Affymetrix HT Rat Focus Array | 1382048_at | - | - |
| Affymetrix HT Rat Focus Array | 1374431_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1100003190.rn.1 | TC1100000609.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374431_at | - | - |
| Rat Expression Set 230 | 1382048_PM_at | - | - |
| Rat Expression Set 230 | 1374431_PM_at | - | - |
| Rat Expression Set 230 | 1382048_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751161 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823609 | - | - |
| Rat Genome U34 Set | rc_AA892006_at | - | - |
| Rat Genome U34 Set | rc_AA893217_at | - | - |
| Rat Genome U34 Set | rc_AA892006_g_at | - | - |
| Rat Genome U34 Set | rc_AI029960_at | - | - |
| Rat Genome U34 Set | rc_AI071788_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21847769 | - | - |

Atp6v1a
Genomic Map