Assay Details
| Gene Symbol: Slc35d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106668.2 | NP_001100138.1 | 10 - 11 | 832 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P012328 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P012325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239410 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849925 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576651 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163523 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P012328 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P012325 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P012325 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P012328 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P012325 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P012328 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P140491 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10878373 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878366 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878363 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878364 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878371 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878367 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878365 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878369 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878372 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878362 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878370 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878368 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10878361 | - | - |
| Rat Genome 230 2.0 Array | 1376187_at | - | - |
| Rat Genome 230 2.0 Array | 1376187_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376187_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376187_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500023614.rn.1 | TC0500003247.rn.1 | RTA TC |
| Rat Expression Set 230 | 1376187_at | - | - |
| Rat Expression Set 230 | 1376187_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878361 | - | - |
| Rat Genome U34 Set | rc_AI177887_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21200143 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357848 | - | - |

Slc35d1
Genomic Map