Assay Details
| Gene Symbol: LOC100911458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228639.2 | XP_006228701.1 | 2 - 3 | 1102 | 88 | |
| Gene Symbol: Papl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759286.1 | XP_008757508.1 | 2 - 3 | 225 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911458
Genomic Map