Assay Details
Gene Symbol: Zfp61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017512.2 | NP_001017512.2 | 1 - 2 | 135 | 64 | |
XM_008758962.1 | XP_008757184.1 | 1 - 2 | 427 | 64 | ||
GenBank mRNA | BC079015.1 | - | 1 - 2 | 82 | 7113620 | 64 |
CB585199.1 | - | 1 - 2 | 135 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC079015.1, NM_001017512.2, XM_008758962.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
