Assay Details
| Gene Symbol: Kirrel2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271398.1 | NP_001258327.1 | 2 - 3 | 872 | 78 | |
| XM_008759099.1 | XP_008757321.1 | 3 - 4 | 828 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kirrel2
Genomic Map