Assay Details
| Gene Symbol: Siglec10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106250.1 | NP_001099720.1 | 3 - 4 | 574 | 138 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106250.1 transcripts.
Set Membership: |
|

Siglec10
Genomic Map