Assay Details
| Gene Symbol: Siglec10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106250.1 | NP_001099720.1 | 7 - 8 | 1720 | 79 | |
| XM_006228948.2 | XP_006229010.1 | 10 - 11 | 1879 | 79 | ||
| GenBank mRNA | BC168854.1 | - | 5 - 6 | 953 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Siglec10
Genomic Map