Assay Details
Gene Symbol: Psmd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100831.1 | NP_001094301.1 | 1 - 2 | 401 | 88 | |
GenBank mRNA | BC105894.1 | - | 1 - 2 | 166 | 88 | |
CD371338.1 | - | 1 - 2 | 411 | 88 | ||
FQ213223.1 | - | 1 - 2 | 201 | 88 | ||
FQ214314.1 | - | 1 - 2 | 198 | 88 | ||
FQ223219.1 | - | 1 - 2 | 244 | 88 | ||
FQ233693.1 | - | 1 - 2 | 232 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
