Assay Details
| Gene Symbol: Dpy19l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135835.2 | NP_001129307.2 | 8 - 9 | 1086 | 64 | |
| XM_006228888.2 | XP_006228950.1 | 8 - 9 | 1578 | 64 | ||
| XM_006228889.2 | XP_006228951.1 | 8 - 9 | 988 | 64 | ||
| XM_006228890.2 | XP_006228952.1 | 8 - 9 | 1665 | 64 | ||
| XM_008759162.1 | XP_008757384.1 | 6 - 7 | 692 | 64 | ||
| XM_008759163.1 | XP_008757385.1 | 6 - 7 | 638 | 64 | ||
| XM_008759164.1 | XP_008757386.1 | 5 - 6 | 607 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dpy19l3
Genomic Map