Assay Details
| Gene Symbol: Chst8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107504.2 | NP_001100974.1 | 2 - 3 | 535 | 100 | |
| XM_006228853.2 | XP_006228915.1 | 2 - 3 | 1420 | 100 | ||
| XM_006228854.2 | XP_006228916.1 | 2 - 3 | 846 | 100 | ||
| GenBank mRNA | BC161905.1 | - | 2 - 3 | 535 | 8367390 | 100 |
More Information
Updated Mapping Information
Set Membership: |
|

Chst8
Genomic Map