Assay Details
Gene Symbol: Chst8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107504.2 | NP_001100974.1 | 3 - 4 | 577 | 70 | |
XM_006228853.2 | XP_006228915.1 | 3 - 4 | 1462 | 70 | ||
XM_006228854.2 | XP_006228916.1 | 3 - 4 | 888 | 70 | ||
GenBank mRNA | BC161905.1 | - | 3 - 4 | 577 | 8367390 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P062670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801864 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133507 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452979 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122582 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34927 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P062670 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P062670 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P062670 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P431169 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1460 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73448 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2102 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10721049 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10721047 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10721048 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10721046 | - | - |
Rat Genome 230 2.0 Array | 1394334_at | - | - |
Rat Genome 230 2.0 Array | 1381897_at | - | - |
Rat Genome 230 2.0 Array | 1381897_PM_at | - | - |
Rat Genome 230 2.0 Array | 1394334_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1394334_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1381897_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1381897_at | - | - |
Affymetrix HT Rat Focus Array | 1394334_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100042150.rn.1 | TC0100005209.rn.1 | RTA TC |
Rat Expression Set 230 | 1394334_PM_at | - | - |
Rat Expression Set 230 | 1381897_PM_at | - | - |
Rat Expression Set 230 | 1394334_at | - | - |
Rat Expression Set 230 | 1381897_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10721046 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21436194 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374579 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353250 | - | - |
