Assay Details
| Gene Symbol: Slc7a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012100.1 | NP_001012100.1 | 2 - 3 | 872 | 89 | |
| GenBank mRNA | BC085796.1 | - | 2 - 3 | 872 | 7109760 | 89 |
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a13
Genomic Map