Assay Details
Gene Symbol: Tstd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_575783.6 | XP_575783.3 | 1 - 2 | 169 | 62 | |
GenBank mRNA | BC078951.1 | - | 1 - 2 | 137 | 62 | |
BC089864.1 | - | 1 - 2 | 149 | 62 | ||
BC104709.1 | - | 1 - 2 | 52 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
