Assay Details
Gene Symbol: RGD1304595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106661.1 | NP_001100131.1 | 9 - 10 | 1228 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P106291 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P103114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159051 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P103114 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P106291 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P106291 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P103114 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P106291 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P103114 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P103114 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 19482 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10869489 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10869488 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10869496 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10869495 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10869490 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10869493 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10869492 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10869494 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10869491 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10869487 | - | - |
Rat Genome 230 2.0 Array | 1379987_at | - | - |
Rat Genome 230 2.0 Array | 1379987_PM_at | - | - |
Rat Genome 230 2.0 Array | 1391817_at | - | - |
Rat Genome 230 2.0 Array | 1391817_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1379987_at | - | - |
Affymetrix HT Rat Focus Array | 1379987_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500005291.rn.1 | TC0500000783.rn.1 | RTA TC |
Rat Expression Set 230 | 1391817_PM_at | - | - |
Rat Expression Set 230 | 1391817_at | - | - |
Rat Expression Set 230 | 1379987_at | - | - |
Rat Expression Set 230 | 1379987_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10869487 | - | - |
Rat Genome U34 Set | rc_AA901178_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21031812 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365947 | - | - |
