Assay Details
| Gene Symbol: Vapa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031631.2 | NP_113819.3 | 1 - 2 | 317 | 84 | |
| XM_008767384.1 | XP_008765606.1 | 1 - 2 | 870 | 84 | ||
| GenBank mRNA | BC061875.1 | - | 1 - 2 | 317 | 5598274 | 84 |
| FQ228815.1 | - | 1 - 2 | 174 | 84 | ||
| FQ234023.1 | - | 1 - 2 | 74 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vapa
Genomic Map