Assay Details
| Gene Symbol: Vapa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031631.2 | NP_113819.3 | 3 - 4 | 563 | 128 | |
| XM_008767384.1 | XP_008765606.1 | 3 - 4 | 1116 | 128 | ||
| GenBank mRNA | AF086630.1 | - | 3 - 4 | 311 | 128 | |
| BC061875.1 | - | 3 - 4 | 563 | 5598274 | 128 | |
| FQ228815.1 | - | 3 - 4 | 420 | 128 | ||
| FQ234023.1 | - | 3 - 4 | 320 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vapa
Genomic Map