Assay Details
Gene Symbol: Fam188a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106122.1 | NP_001099592.1 | 7 - 8 | 837 | 66 | |
GenBank mRNA | FQ214325.1 | - | 4 - 5 | 508 | 66 | |
FQ214453.1 | - | 2 - 3 | 367 | 66 | ||
FQ230167.1 | - | 1 - 2 | 196 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
