Assay Details
Gene Symbol: St8sia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213624.1 | NP_998789.1 | 723 | 76 | ||
GenBank mRNA | AJ699423.1 | - | 723 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P015467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63371 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161040 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4600 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634450 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932543 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38012 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P015467 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P015467 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P015467 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P543171 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 65057 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23564 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 89883 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 16765 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 39114 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95636 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 70758 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95309 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 39483 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24849 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10799832 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799827 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799828 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799825 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799831 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799830 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799829 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799826 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10799824 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799824 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368410 | - | - |
RatRef-12 Expression BeadChip | ILMN_59345 | - | - |
